Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177092.4 | NP_796066.1 | 2, 3 | 264 | ||
GenBank mRNA | AK086975.1 | - | 2, 3 | 231 | ||
BB617432.1 | - | 2, 3 | 299 | |||
BC096447.1 | - | 2, 3 | 145 | 6310686 |