Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138590.4 | NP_613056.2 | 2 | 313 | ||
GenBank mRNA | AK029808.1 | - | 2 | 320 | ||
AK032353.1 | - | 2 | 285 | |||
AK039158.1 | - | 2 | 234 | |||
AK043238.1 | - | 2 | 226 | |||
AK080785.1 | - | 2 | 226 | |||
AK157728.1 | - | 2 | 216 | |||
BC050200.1 | - | 1 | 188 | |||
BC117020.1 | - | 2 | 216 | 40126339 | ||
BC117022.1 | - | 2 | 216 | 40126341 | ||
BC144774.1 | - | 2 | 220 | 9053322 | ||
BY769121.1 | - | 336 | ||||
CJ150974.1 | - | 2 | 314 | |||
CJ155194.1 | - | 2 | 285 |