Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001412.3 | NP_001403.1 | 3 | 386 | ||
GenBank mRNA | AK312320.1 | - | 1 | 373 | ||
BC000793.1 | - | 3 | 347 | 3451631 | ||
BC067851.1 | - | 3 | 373 | 5288453 | ||
BC110421.1 | - | 3 | 351 | |||
BT007064.1 | - | 1 | 178 | |||
L18960.1 | - | 3 | 385 |