Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177082.4 | NP_796056.2 | 3 | 3321 | ||
GenBank mRNA | AK030745.1 | - | 2 | 3144 | ||
AK053093.1 | - | 3 | 3322 | |||
BC082582.1 | - | 3 | 3013 | 30653287 |