Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015794.1 | NP_056609.1 | 9 | 14392 | ||
GenBank mRNA | AK047946.1 | - | 1 | 593 | ||
AK086201.1 | - | 1 | 624 | |||
AK086431.1 | - | 1 | 401 |