Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025611.5 | NP_079887.3 | 2 | 574 | ||
GenBank mRNA | AK082134.1 | - | 1 | 1265 | ||
AK144856.1 | - | 1 | 1053 | |||
AK147156.1 | - | 2 | 332 | |||
AK147905.1 | - | 2 | 288 | |||
AK155454.1 | - | 1 | 1061 | |||
AK155583.1 | - | 2 | 578 | |||
AK166135.1 | - | 2 | 350 | |||
AK170580.1 | - | 2 | 577 | |||
AK170808.1 | - | 2 | 348 | |||
AK217082.1 | - | 1 | 135 | |||
BC065096.1 | - | 1 | 210 | |||
BC138998.1 | - | 2 | 512 | 8862020 | ||
BC139000.1 | - | 2 | 512 | 8862022 |