Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PASK SIRNA Assays ›
Gene Symbol:
PASK
Gene Name:
PAS domain containing serine/threonine kinase
Gene Aliases:
PASKIN, STK37
Chromosome Location:
Chr.2: 241106099 - 241150347 on Build GRCh38
UniGene ID:
Hs.397891
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
978
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PASK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001252119.1
NP_001239048.1
10
1742
NM_001252120.1
NP_001239049.1
11
1895
NM_001252122.1
NP_001239051.1
10
1637
NM_001252124.1
NP_001239053.1
10
1742
NM_015148.3
NP_055963.2
10
1742
XM_005246991.1
XP_005247048.1
7
1077
XM_011510826.1
XP_011509128.1
10
1697
XM_011510828.1
XP_011509130.1
11
1798
XM_011510829.1
XP_011509131.1
10
1739
XM_011510830.1
XP_011509132.1
10
1740
XM_011510831.2
XP_011509133.1
8
1904
XM_011510832.2
XP_011509134.1
8
1904
XM_011510833.1
XP_011509135.1
7
1077
XM_011510834.1
XP_011509136.1
3
800
XM_011510835.1
XP_011509137.1
3
390
XM_017003634.1
XP_016859123.1
10
1986
XM_017003635.1
XP_016859124.1
11
1798
XM_017003636.1
XP_016859125.1
10
1754
GenBank mRNA
AF387103.1
-
10
1682
AK056516.1
-
10
1687
AK296945.1
-
7
1247
AK302417.1
-
10
1593
AK302561.1
-
7
1077
AL117400.1
-
2
2041
BC020054.1
-
6
917
BC043495.1
-
4
499
BC050565.1
-
11
1701
4537404
BC063585.1
-
10
1685
4419998
CR749231.1
-
11
1896
D50925.2
-
10
1673
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
regulation of respiratory gaseous exchange
negative regulation of glycogen biosynthetic process
positive regulation of translation
protein autophosphorylation
regulation of glucagon secretion
regulation of energy homeostasis
protein serine/threonine kinase activity
protein binding
ATP binding
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0