Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177190.5 | NP_796164.1 | 3 | 428 | ||
GenBank mRNA | AK047525.1 | - | 2 | 254 | ||
AK082024.1 | - | 3 | 430 | |||
BC116926.1 | - | 2 | 249 | 40126245 | ||
BC116932.1 | - | 2 | 249 | 40126251 |