Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172925.2 | NP_766513.2 | 3 | 4897 | ||
GenBank mRNA | AK036669.1 | - | 1 | 1436 | ||
AK085911.1 | - | 1 | 2101 | |||
BC094497.1 | - | 1 | 833 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177238.1 | - | 2101 | Chr. - - [-] on Build |