Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029851.2 | NP_084127.2 | 1 | 211 | ||
GenBank mRNA | AK032860.1 | - | 1 | 185 | ||
AK045978.1 | - | 1 | 157 | |||
AK046538.1 | - | 1 | 210 | |||
AK084796.1 | - | 1 | 175 | |||
AY452064.1 | - | 503 | ||||
BC088991.1 | - | 1 | 130 | |||
DQ104402.1 | - | 1 | 211 |