Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177307.3 | NP_796281.1 | 4 | 426 | ||
GenBank mRNA | AK028950.1 | - | 4 | 427 | ||
AK037192.1 | - | 4 | 427 | |||
BC145755.1 | - | 4 | 423 | 40131058 |