Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177323.3 | NP_796297.2 | 3 | 291 | ||
GenBank mRNA | AK028707.1 | - | 3 | 322 | ||
AK036805.1 | - | 3 | 291 | |||
AK133789.1 | - | 3 | 317 | |||
AK135334.1 | - | 3 | 312 | |||
AK136479.1 | - | 3 | 297 | |||
AK144144.1 | - | 3 | 313 | |||
BC065059.1 | - | 3 | 291 | 6401664 | ||
BC145267.1 | - | 3 | 281 | 9053821 | ||
BC150745.1 | - | 3 | 281 | 9087656 |