Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPAGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001382.3 | NP_001373.2 | 2 | 680 | ||
XM_005271422.2 | XP_005271479.1 | 2 | 696 | |||
XM_017017294.1 | XP_016872783.1 | 2 | 696 | |||
GenBank mRNA | AK095718.1 | - | 1 | 1103 | ||
AK128572.1 | - | 4 | 1812 | |||
AK225994.1 | - | 2 | 425 | |||
AK312783.1 | - | 2 | 483 | |||
BC000325.2 | - | 2 | 428 | 2821845 | ||
BT006802.1 | - | 2 | 260 | |||
Z82022.1 | - | 2 | 339 |