Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177327.5 | NP_796301.2 | 7, 8 | 830 | ||
GenBank mRNA | AK032232.1 | - | 7, 8 | 821 | ||
AK137588.1 | - | 7, 8 | 755 | |||
AK168567.1 | - | 7, 8 | 781 | |||
BC150812.1 | - | 6, 7 | 665 | 9087723 |