Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001388.4 | NP_001379.1 | 2 | 324 | ||
XM_005256499.2 | XP_005256556.1 | 2 | 306 | |||
XM_005256500.2 | XP_005256557.1 | 2 | 306 | |||
XM_011523704.1 | XP_011522006.1 | 2 | 306 | |||
XM_011523705.1 | XP_011522007.1 | 2 | 306 | |||
XM_011523706.1 | XP_011522008.1 | 2 | 219 | |||
XM_011523707.1 | XP_011522009.1 | 2 | 270 | |||
XM_017024296.1 | XP_016879785.1 | 2 | 219 | |||
XM_017024297.1 | XP_016879786.1 | 2 | 270 | |||
GenBank mRNA | AK295575.1 | - | 2 | 212 | ||
AK297569.1 | - | 2 | 219 | |||
AK313362.1 | - | 2 | 243 | |||
BC000493.2 | - | 2 | 222 | 2820542 | ||
BT006976.1 | - | 2 | 167 | |||
DC298961.1 | - | 2 | 324 | |||
X80754.1 | - | 2 | 214 |