Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177323.3 | NP_796297.2 | 15 | 2496 | ||
GenBank mRNA | AK028707.1 | - | 14 | 2353 | ||
AK135334.1 | - | 15 | 2517 | |||
BC065059.1 | - | 15 | 2496 | 6401664 |