Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb2b24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177446.2 | NP_803229.1 | 2 | 226 | ||
GenBank mRNA | AA794526.1 | - | 2 | 226 | ||
AY370635.1 | - | 2 | 243 | |||
BC115602.1 | - | 2 | 226 | 40102624 | ||
BC115603.1 | - | 2 | 226 | 40102625 | ||
GU269236.1 | - | 2 | 216 |