Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hcc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177464.4 | NP_803415.1 | 7 | 3070 | ||
GenBank mRNA | AK077281.1 | - | 4 | 1033 | ||
AK082926.1 | - | 7 | 3071 | |||
AK147893.1 | - | 7 | 3054 | |||
BC038935.1 | - | 7 | 3047 | 5035077 |