Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umodl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177465.4 | NP_803416.2 | 22 | 4878 | ||
GenBank mRNA | AK040446.1 | - | 14 | 3379 | ||
AY771616.1 | - | 24 | 4757 | |||
AY771617.1 | - | 23 | 4844 | |||
AY771618.1 | - | 23 | 4499 | |||
BC051427.1 | - | 22 | 4533 |