Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199677.1 | NP_001186606.1 | 4 | 1414 | ||
NM_177574.4 | NP_808242.1 | 2 | 1159 | |||
GenBank mRNA | AK090181.1 | - | 2 | 1161 | ||
AK158287.1 | - | 4 | 1343 | |||
AY081953.2 | - | 4 | 1414 | |||
AY352519.1 | - | 2 | 1159 | |||
BC044789.1 | - | 4 | 1317 | 3484221 | ||
BU704550.1 | - | 1 | 270 |