Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177602.3 | NP_808270.2 | 6, 7 | 1450 | ||
GenBank mRNA | AK081117.1 | - | 3, 4 | 398 | ||
AK147993.1 | - | 7, 8 | 3152 |