Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L2 SIRNA Assays ›
Gene Symbol:
EPB41L2
Gene Name:
erythrocyte membrane protein band 4.1 like 2
Gene Aliases:
4.1-G, 4.1G
Chromosome Location:
Chr.6: 130829111 - 131063322 on Build GRCh38
UniGene ID:
Hs.486470
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9864
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135554.1
NP_001129026.1
2
319
NM_001135555.3
NP_001129027.1
2
332
NM_001199388.2
NP_001186317.1
2
332
NM_001252660.1
NP_001239589.1
2
299
NM_001431.3
NP_001422.1
2
319
XM_006715362.2
XP_006715425.1
2
350
XM_011535524.1
XP_011533826.1
2
319
XM_011535525.1
XP_011533827.1
2
319
XM_011535527.1
XP_011533829.1
2
319
XM_011535528.1
XP_011533830.1
2
319
XM_011535529.1
XP_011533831.1
2
319
XM_011535530.1
XP_011533832.1
2
319
XM_011535534.1
XP_011533836.1
2
319
XM_011535536.1
XP_011533838.1
2
319
XM_017010349.1
XP_016865838.1
2
347
XM_017010350.1
XP_016865839.1
2
281
XM_017010351.1
XP_016865840.1
2
343
XM_017010352.1
XP_016865841.1
2
185
XM_017010353.1
XP_016865842.1
2
319
XM_017010354.1
XP_016865843.1
2
319
XM_017010355.1
XP_016865844.1
2
347
XM_017010356.1
XP_016865845.1
2
319
XM_017010357.1
XP_016865846.1
2
281
XM_017010358.1
XP_016865847.1
2
319
XM_017010359.1
XP_016865848.1
2
347
XM_017010360.1
XP_016865849.1
2
185
XM_017010361.1
XP_016865850.1
2
347
XM_017010362.1
XP_016865851.1
2
281
XM_017010363.1
XP_016865852.1
2
309
GenBank mRNA
AB209522.1
-
2
230
AF027299.1
-
2
181
AK295124.1
-
2
261
AK307764.1
-
2
352
AL698619.1
-
2
243
AY047584.1
-
1
145
BI461477.1
-
2
293
CR749262.1
-
2
261
CR936793.1
-
2
299
DA108957.1
-
2
319
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cortical actin cytoskeleton organization
actin binding
structural molecule activity
spectrin binding
PH domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0