Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280.2 | NP_001271.1 | 7 | 894 | ||
NM_001300829.1 | NP_001287758.1 | 6 | 1271 | |||
NR_023312.2 | 6 | 1330 | ||||
NR_023313.2 | 5 | 1162 | ||||
XM_006722637.1 | XP_006722700.1 | 7 | 957 | |||
GenBank mRNA | AF021336.1 | - | 7 | 827 | ||
AK094781.1 | - | 5 | 1095 | |||
AK095781.1 | - | 4 | 1389 | |||
AK128423.1 | - | 5 | 1184 | |||
AK304185.1 | - | 5 | 1348 | |||
AK304470.1 | - | 6 | 1103 | |||
BC000403.2 | - | 7 | 806 | 2822968 | ||
BC000901.1 | - | 7 | 811 | 2900585 | ||
D78134.1 | - | 7 | 825 |