Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199775.1 | NP_001186704.1 | 21 | 7000 | ||
NM_001304.4 | NP_001295.2 | 21 | 7638 | |||
GenBank mRNA | AL049339.1 | - | 1 | 873 | ||
BC020457.1 | - | 1 | 1137 |