Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177769.4 | NP_808437.2 | 7 | 866 | ||
GenBank mRNA | AK029207.1 | - | 7 | 747 | ||
AK132239.1 | - | 7 | 867 | |||
AK164094.1 | - | 18 | ||||
BC120566.1 | - | 6 | 582 | |||
BC144961.1 | - | 6 | 582 | |||
BC144963.1 | - | 6 | 544 | 9053511 |