Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177699.4 | NP_808367.2 | 20 | 3353 | ||
GenBank mRNA | AB041045.1 | - | 20 | 3247 | ||
AK030737.1 | - | 20 | 3315 | |||
BC017144.1 | - | 17 | 3784 | |||
BC060654.1 | - | 20 | 3336 | 30354865 |