Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177715.4 | NP_808383.3 | 1 | 2973 | ||
GenBank mRNA | AK032276.1 | - | 1 | 858 | ||
AK035535.1 | - | 1 | 2974 | |||
AK049420.1 | - | 1 | 1098 | |||
AK086988.1 | - | 1 | 845 | |||
AK172581.1 | - | 1 | 2973 | |||
AK173260.1 | - | 1 | 2478 | |||
BC116647.1 | - | 2 | 2360 | |||
BC116648.1 | - | 2 | 2361 |