Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nalcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177393.4 | NP_796367.3 | 44 | 5457 | ||
GenBank mRNA | AK044431.1 | - | 12 | 1404 |