Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025426.2 | NP_001020597.1 | 9 | 927 | ||
NM_001170567.1 | NP_001164038.1 | 8 | 624 | |||
NM_177786.4 | NP_808454.2 | 8 | 624 | |||
GenBank mRNA | AK043137.1 | - | 8 | 576 | ||
AK052458.1 | - | 8 | 612 | |||
AK078771.1 | - | 8 | 569 | |||
AK136321.1 | - | 8 | 569 | |||
AK166366.1 | - | 9 | 890 | |||
BC072604.1 | - | 9 | 927 | 6831032 |