siRNA Details
Gene Symbol: Zfp459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177811.4 | NP_808479.1 | 4 | 427 | ||
GenBank mRNA | AK036923.1 | - | 4 | 428 | ||
BC100389.1 | - | 4 | 393 |
Gene Symbol: Rsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013769.1 | NP_001013791.1 | 4 | 422 | ||
GenBank mRNA | AK087838.1 | - | 1 | 46 | ||
BC103788.1 | - | 4 | 397 | 6333828 |
Gene Symbol: Zfp429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080941.1 | NP_001074410.1 | 4 | 422 | ||
GenBank mRNA | AK083314.1 | - | 5 | 490 | ||
BC046484.1 | - | 4 | 406 |
Gene Symbol: Zfp455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048204.1 | NP_001041669.1 | 4 | 439 | ||
GenBank mRNA | AK028336.1 | - | 3 | 284 |
Gene Symbol: Zfp456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001186.3 | NP_001001186.3 | 4 | 424 | ||
GenBank mRNA | AK164227.1 | - | 4 | 425 | ||
BB654999.1 | - | 4 | 425 | |||
BC141197.1 | - | 4 | 407 | 9055809 | ||
BY723566.1 | - | 4 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
