Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001361.4 | NP_001352.2 | 6 | 815 | ||
XM_005255827.3 | XP_005255884.1 | 6 | 1120 | |||
XM_005255829.3 | XP_005255886.1 | 5 | 757 | |||
XM_017022990.1 | XP_016878479.1 | 6 | 895 | |||
GenBank mRNA | AK292293.1 | - | 7 | 971 | ||
BC065245.1 | - | 6 | 795 | 6064723 | ||
M94065.1 | - | 7 | 797 |