Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198811.1 | NP_001185740.1 | 20 | 4161 | ||
NM_177863.4 | NP_808531.2 | 19 | 3951 | |||
GenBank mRNA | AB160986.1 | - | 20 | 4161 | ||
AJ616838.1 | - | 19 | 3951 | |||
AK052629.1 | - | 3 | 412 | |||
BC138427.1 | - | 19 | 3862 | 8861447 | ||
BC145274.1 | - | 19 | 3862 |