Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025163.1 | NP_001020334.1 | 3 | 213 | ||
NM_001112805.1 | NP_001106276.1 | 3 | 213 | |||
NM_177888.4 | NP_808556.1 | 4 | 452 | |||
GenBank mRNA | AK039658.1 | - | 4 | 425 | ||
AK134974.1 | - | 3 | 242 | |||
AK163133.1 | - | 3 | 212 | |||
AK173164.1 | - | 3 | 233 | |||
BB603663.2 | - | 3 | 214 | |||
BC085322.1 | - | 3 | 204 | 30739758 | ||
BY768295.1 | - | 3 | 214 |