Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177845.4 | NP_808513.2 | 11 | 1483 | ||
GenBank mRNA | AB195277.1 | - | 11 | 1386 | ||
AK049063.1 | - | 10 | 2016 | |||
BC060600.1 | - | 8 | 1429 | |||
BC066085.1 | - | 11 | 1480 | |||
BC137899.1 | - | 11 | 1260 | 8860918 | ||
BC145020.1 | - | 12 | 1371 |