Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177911.4 | NP_808579.2 | 14 | 2424 | ||
GenBank mRNA | AK035279.1 | - | 14 | 2425 |