Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289670.1 | NP_001276599.1 | 7, 8 | 761 | ||
NM_001289682.1 | NP_001276611.1 | 8, 9 | 844 | |||
NM_178252.3 | NP_839983.1 | 8, 9 | 871 | |||
GenBank mRNA | AK142180.1 | - | 7, 8 | 762 | ||
AY217764.1 | - | 7, 8 | 704 | |||
BC061471.1 | - | 1, 2 | 48 | 5698780 | ||
BC065086.1 | - | 8, 9 | 820 | 6850475 | ||
BC065166.1 | - | 8, 9 | 818 | 6836510 | ||
BC066047.1 | - | 8, 9 | 796 | 6822191 |