Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178440.4 | NP_848534.2 | 1 | 199 | ||
GenBank mRNA | AK030825.1 | - | 1 | 200 | ||
AK036461.1 | - | 1 | 184 | |||
AK042033.1 | - | 1 | 150 | |||
AK088011.1 | - | 1 | 200 | |||
AK146251.1 | - | 1 | 172 | |||
AK152797.1 | - | 1 | 193 | |||
AK159700.1 | - | 1 | 175 | |||
AK169450.1 | - | 1 | 193 | |||
AK171284.1 | - | 1 | 193 | |||
AK171505.1 | - | 1 | 172 | |||
BC113755.1 | - | 1 | 143 | 40061202 |