Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178240.2 | NP_839971.1 | 1 | 320 | ||
GenBank mRNA | AY081947.1 | - | 1 | 320 | ||
BC130022.1 | - | 1 | 323 | 40140742 |