Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290633.1 | NP_001277562.1 | 5, 6 | 870 | ||
NM_178256.4 | NP_839987.2 | 5, 6 | 873 | |||
GenBank mRNA | AK147418.1 | - | 5, 6 | 521 | ||
AK147432.1 | - | 2, 3 | 152 | |||
AY263368.1 | - | 5, 6 | 818 | |||
BC139012.1 | - | 5, 6 | 749 | 8862034 | ||
BC145500.1 | - | 5, 6 | 746 | 9054063 |