Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930486L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178098.2 | NP_835199.1 | 4 | 435 | ||
GenBank mRNA | AK015635.1 | - | 4 | 405 | ||
AK145933.1 | - | 4 | 436 | |||
AY146988.1 | - | 4 | 435 | |||
BC061218.1 | - | 4 | 422 | 30281034 |