Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289656.1 | NP_001276585.1 | 5 | 1079 | ||
NM_001289657.1 | NP_001276586.1 | 5 | 1079 | |||
NM_178630.4 | NP_848745.2 | 5 | 1079 | |||
GenBank mRNA | AK029793.1 | - | 5 | 1080 | ||
AK029798.1 | - | 5 | 663 | |||
AK030378.1 | - | 5 | 637 | |||
AK047649.1 | - | 5 | 636 | |||
AK054066.1 | - | 4 | 2110 | |||
BC139065.1 | - | 5 | 531 | 8862087 | ||
DQ867031.1 | - | 5 | 1080 | |||
DQ867032.1 | - | 5 | 1080 |