Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luzp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178705.5 | NP_848820.4 | 2 | 788 | ||
GenBank mRNA | AK032204.1 | - | 2 | 445 | ||
AK034077.1 | - | 2 | 406 | |||
AK082126.1 | - | 2 | 536 | |||
AY164459.1 | - | 2 | 785 | |||
BC075646.1 | - | 2 | 418 | 30620475 |