Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145977.1 | NP_001139449.1 | 2 | 755 | ||
NM_178721.4 | NP_848836.2 | 3 | 782 | |||
GenBank mRNA | AK033973.1 | - | 2 | 382 | ||
AK038842.1 | - | 3 | 761 | |||
AK043760.1 | - | 3 | 765 | |||
AK046800.1 | - | 2 | 658 | |||
AK163636.1 | - | 3 | 759 | |||
AK165030.1 | - | 3 | 779 | |||
BC139401.1 | - | 3 | 351 | 8862423 | ||
BC139402.1 | - | 3 | 351 | 8862424 |