Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172121.1 | NP_001165592.1 | 4 | 476 | ||
NM_001172122.1 | NP_001165593.1 | 3 | 948 | |||
NM_001172123.1 | NP_001165594.1 | 3 | 948 | |||
NM_001172124.1 | NP_001165595.1 | 3 | 948 | |||
NM_001172126.1 | NP_001165597.1 | 3 | 948 | |||
NM_178660.4 | NP_848775.2 | 3 | 948 | |||
GenBank mRNA | AK030478.1 | - | 3 | 608 | ||
AK030609.1 | - | 3 | 582 | |||
AK032938.1 | - | 3 | 945 | |||
AK033005.1 | - | 3 | 591 | |||
AK041098.1 | - | 3 | 950 | |||
AK046197.1 | - | 4 | 477 | |||
AK051776.1 | - | 3 | 947 | |||
AK052196.1 | - | 3 | 929 | |||
AK081921.1 | - | 3 | 942 | |||
AK162457.1 | - | 3 | 313 | |||
BC108402.1 | - | 3 | 922 | 6516420 | ||
BC117841.1 | - | 3 | 388 | 40093225 |