Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Themis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB509340.1 | - | 1, 2 | 184 | ||
AK040594.1 | - | 1, 2 | 363 | |||
AK041386.1 | - | 1, 2 | 400 | |||
AK041562.1 | - | 1, 2 | 289 | |||
AK041622.1 | - | 1, 2 | 363 | |||
AK088136.1 | - | 1, 2 | 365 | |||
AK089077.1 | - | 1, 2 | 365 | |||
AK138701.1 | - | 1, 2 | 363 | |||
AK169558.1 | - | 1, 2 | 133 | |||
BB638055.1 | - | 1, 2 | 400 | |||
BC094898.1 | - | 126 | ||||
BC125486.1 | - | 1, 2 | 246 | 40130001 | ||
BC125488.1 | - | 1, 2 | 246 | 40130003 |