Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178715.3 | NP_848830.1 | 1 | 1344 | ||
GenBank mRNA | AK030302.1 | - | 1 | 1339 | ||
AK033580.1 | - | 1 | 1345 | |||
AK037246.1 | - | 1 | 1342 | |||
AK169014.1 | - | 1 | 1380 | |||
BC119224.1 | - | 1 | 1337 | 8733973 | ||
BC119226.1 | - | 1 | 1337 | 8733975 |