Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289656.1 | NP_001276585.1 | 9 | 2151 | ||
NM_001289657.1 | NP_001276586.1 | 8 | 2125 | |||
NM_178630.4 | NP_848745.2 | 8 | 2136 | |||
GenBank mRNA | AK029793.1 | - | 9 | 2152 | ||
AK029798.1 | - | 8 | 1709 | |||
AK030378.1 | - | 8 | 1694 | |||
AK054066.1 | - | 7 | 3156 | |||
BC139065.1 | - | 8 | 1588 | 8862087 | ||
DQ867031.1 | - | 8 | 2137 | |||
DQ867032.1 | - | 8 | 2137 |