Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242349.1 | NP_001229278.1 | 17 | 2016 | ||
NM_178642.5 | NP_848757.4 | 18 | 2025 | |||
GenBank mRNA | AK029329.1 | - | 4 | 392 | ||
AK032793.1 | - | 13 | 1037 | |||
AK052589.1 | - | 18 | 1977 | |||
BC062959.1 | - | 17 | 1902 | 30547439 |