Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178691.4 | NP_848806.2 | 2 | 3330 | ||
GenBank mRNA | AK036938.1 | - | 2 | 3332 | ||
AK036991.1 | - | 2 | 3333 | |||
AK044678.1 | - | 2 | 3130 | |||
AK053730.1 | - | 2 | 3119 | |||
BC099948.1 | - | 2 | 3123 | 30097649 |