Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025091.1 | NP_001020262.1 | 14 | 1428 | ||
NM_001090.2 | NP_001081.1 | 13 | 1314 | |||
GenBank mRNA | AB209564.1 | - | 6 | 591 | ||
AF027302.1 | - | 13 | 1313 | |||
AK298061.1 | - | 15 | 1305 | |||
AK307551.1 | - | 14 | 1382 | |||
AL832430.1 | - | 8 | 858 | |||
BC034488.2 | - | 14 | 1373 | 4826422 | ||
BC112923.1 | - | 14 | 1361 | 7470309 |